Learn More
Biolegend Purified anti-human CD45RB
Supplier:  Biolegend 310202
Encompass_Preferred
Purified anti-human CD45RB [MEM-55]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IHC-P, IP, WB; Size: 100 ug
Catalog No. 50-166-540