Learn More
Biolegend Purified anti-human CD45RO
Supplier:  Biolegend 304202
Encompass_Preferred
Purified anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC, WB, IP, IHC-P; Size: 100 ug
Catalog No. 50-166-093