Learn More
Biolegend Purified anti-human CD5
Supplier:  Biolegend 300602
Encompass_Preferred
Purified anti-human CD5 [UCHT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Capuchin Monkey, Common Marmoset, Cynomolgus, Rhesus; Apps: FC, IHC-F, WB; Size: 100 ug
Catalog No. 50-165-604