Learn More
Biolegend Purified anti-human CD5 (Maxpar Ready)
Supplier:  Biolegend 300627
Encompass_Preferred
Purified anti-human CD5 (Maxpar Ready) [UCHT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Capuchin Monkey, Common Marmoset, Cynomolgus, Rhesus; Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-207-2704