Learn More
Biolegend Purified anti-human CD56 (NCAM)
Supplier:  Biolegend 304601
Encompass_Preferred
Purified anti-human CD56 (NCAM) [MEM-188]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cattle (Bovine, Cow); Apps: FC, IHC-P, IP, WB; Size: 25 ug
Catalog No. 50-166-133