Learn More
Biolegend Purified anti-human CD58 (LFA-3)
Supplier:  Biolegend 330902
Encompass_Preferred
Purified anti-human CD58 (LFA-3) [TS2/9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IP; Size: 100 ug
Catalog No. 50-167-600