Learn More
Biolegend Purified anti-human CD59
Supplier:  Biolegend 304702
Encompass_Preferred
Purified anti-human CD59 [p282 (H19)]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 ug
Catalog No. 50-166-152