Learn More
Biolegend Purified anti-human CD61
Supplier:  Biolegend 336402
Encompass_Preferred
Purified anti-human CD61 [VI-PL2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, CyTOF, ICC, IHC-F, WB; Size: 100 ug
Catalog No. 50-167-860