Learn More
Biolegend Purified anti-human CD62L
Supplier:  Biolegend 304802
Encompass_Preferred
Purified anti-human CD62L [DREG-56]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cattle (Bovine, Cow); Apps: FC, WB, Block; Size: 100 ug
Catalog No. 50-166-156