Learn More
Biolegend Purified anti-human CD62L (Maxpar Ready)
Supplier:  Biolegend 304835
Encompass_Preferred
Purified anti-human CD62L (Maxpar Ready) [DREG-56]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cattle (Bovine, Cow); Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-850-540