Learn More
Biolegend Purified anti-human CD84
Supplier:  Biolegend 326002
Encompass_Preferred
Purified anti-human CD84 [CD84.1.21]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IP; Size: 100 ug
Catalog No. 50-167-308