Learn More
Biolegend Purified anti-human CD8a
Supplier:  Biolegend 300902
Encompass_Preferred
Purified anti-human CD8a [HIT8a]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Marmoset; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-165-623