Learn More
Biolegend Purified anti-human CD9
Supplier:  Biolegend 312102
Encompass_Preferred
Purified anti-human CD9 [HI9a]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Rhesus, Sheep (Ovine); Apps: FC, ICC; Size: 100 ug
Catalog No. 50-605-256