Learn More
Biolegend Purified anti-human CD94
Supplier:  Biolegend 305502
Encompass_Preferred
Purified anti-human CD94 [DX22]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-166-270