Learn More
Biolegend Purified anti-human CRP (C-reactive protein)
Supplier:  Biolegend 696006
Encompass_Preferred
Purified anti-human CRP (C-reactive protein) [A16065C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-208-7670