Learn More
Biolegend Purified anti-human CD152 (CTLA-4)
Supplier:  Biolegend 525402
Encompass_Preferred
Purified anti-human CD152 (CTLA-4) [A3.6B10.G1]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-605-565