Learn More
Biolegend Purified anti-human CXCL10 (IP-10)
Supplier:  Biolegend 519501
Encompass_Preferred
Purified anti-human CXCL10 (IP-10) [J034D6]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: ICFC, WB; Size: 50 ug
Catalog No. 50-208-5918