Learn More
Biolegend Purified anti-human G-CSF
Supplier:  Biolegend 502101
Encompass_Preferred
Purified anti-human G-CSF [BVD13-3A5]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IHC; Size: 50 ug
Catalog No. 50-169-710