Learn More
Biolegend Purified anti-human GM-CSF
Supplier:  Biolegend 502301
Encompass_Preferred
Purified anti-human GM-CSF [BVD2-21C11]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Rhesus, Cynomolgus; Apps: ELISA Capture, ICFC, IHC, IP, WB; Size: 50 ug
Catalog No. 50-169-717