Learn More
Biolegend Purified anti-human GM-CSF
Supplier:  Biolegend 502201
Encompass_Preferred
Purified anti-human GM-CSF [BVD2-23B6]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Capture, IP, WB; Size: 50 ug
Catalog No. 50-169-713