Learn More
Biolegend Purified anti-human Granzyme A
Supplier:  Biolegend 507202
Encompass_Preferred
Purified anti-human Granzyme A [CB9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC, ICC, IP, IHC-P; Size: 100 ug
Catalog No. 50-170-014