Learn More
Biolegend Purified anti-human IFN-γ
Supplier:  Biolegend 502402
Encompass_Preferred
Purified anti-human IFN-γ [NIB42]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-169-723