Learn More
Biolegend Purified anti-human IL-1 RA
Supplier:  Biolegend 509901
Encompass_Preferred
Purified anti-human IL-1 RA [JK1RA-1]; Isotype: Mouse IgG1; Reactivity: Human; Apps: ELISA Capture; Size: 50 ug
Catalog No. 50-170-070