Learn More
Biolegend Purified anti-human IL-10
Supplier:  Biolegend 501401
Encompass_Preferred
Purified anti-human IL-10 [JES3-9D7]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Rhesus, Cynomolgus; Apps: ELISA Capture, IHC-P, Neut; Size: 50 ug
Catalog No. 50-169-671