Learn More
Biolegend Purified anti-human IL-10
Supplier:  Biolegend 501505
Encompass_Preferred
Purified anti-human IL-10 [JES3-12G8]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA, ICFC, IHC; Size: 50 ug
Catalog No. 50-169-689