Learn More
Biolegend Purified anti-human IL-10
Supplier:  Biolegend 506801
Encompass_Preferred
Purified anti-human IL-10 [JES3-19F1]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Capture, IHC-F, ICC, WB; Size: 50 ug
Catalog No. 50-169-986