Learn More
Biolegend Purified anti-human IL-12/IL-23 p40
Supplier:  Biolegend 501801
Encompass_Preferred
Purified anti-human IL-12/IL-23 p40 [C11.5]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICFC, IHC-F, ICC, Block, Neut, WB; Size: 50 ug
Catalog No. 50-169-692