Learn More
Biolegend Purified anti-human IL-12 (p70)
Supplier:  Biolegend 511001
Encompass_Preferred
Purified anti-human IL-12 (p70) [7B12]; Isotype: Rat IgG1; Reactivity: Human, p70 heterodimer; Apps: ELISA Capture; Size: 50 ug
Catalog No. 50-170-084