Learn More
Biolegend Purified anti-human IL-15
Supplier:  Biolegend 515001
Encompass_Preferred
Purified anti-human IL-15 [BH1509]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture; Size: 50 ug
Catalog No. 50-208-5881