Learn More
Biolegend Purified anti-human IL-1α
Supplier:  Biolegend 500104
Encompass_Preferred
Purified anti-human IL-1α [364-3B3-14]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: ELISA Capture, IHC; Size: 500 ug
Catalog No. 50-169-587