Learn More
Biolegend Purified anti-human IL-2 (Maxpar Ready)
Supplier:  Biolegend 500339
Encompass_Preferred
Purified anti-human IL-2 (Maxpar Ready) [MQ1-17H12]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cat (Feline), Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Sooty Mangabey; Apps: ELISA Capture, CyTOF; Size: 100 ug
Catalog No. 50-208-5484