Learn More
Biolegend Purified anti-human IL-32αβδ
Supplier:  Biolegend 513601
Encompass_Preferred
Purified anti-human IL-32αβδ [KU32-56]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, WB, IF, IP; Size: 50 ug
Catalog No. 50-170-162