Learn More
Biolegend Purified anti-human IL-4
Supplier:  Biolegend 500802
Encompass_Preferred
Purified anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine), Rhesus; Apps: ELISA, CyTOF, Neut, ICFC, IHC; Size: 500 ug
Catalog No. 50-169-633