Learn More
Biolegend Purified anti-human IL-5
Supplier:  Biolegend 500902
Encompass_Preferred
Purified anti-human IL-5 [JES1-39D10]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: ELISA Capture, IHC-F, ICC, WB; Size: 500 ug
Catalog No. 50-169-648