Learn More
Biolegend Purified anti-human IL-7
Supplier:  Biolegend 501302
Encompass_Preferred
Purified anti-human IL-7 [BVD10-40F6]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-169-668