Learn More
Biolegend Purified anti-human IL-9
Supplier:  Biolegend 507602
Encompass_Preferred
Purified anti-human IL-9 [MH9A4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-170-029