Learn More
Biolegend Purified anti-human Jagged 2
Supplier:  Biolegend 346902
Encompass_Preferred
Purified anti-human Jagged 2 [MHJ2-523]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-168-398