Learn More
Biolegend Purified anti-human Lymphotoxin β Receptor (LT-βR)
Supplier:  Biolegend 322002
Encompass_Preferred
Purified anti-human Lymphotoxin β Receptor (LT-βR) [31G4D8]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC, IP, WB; Size: 100 ug
Catalog No. 50-167-105