Learn More
Biolegend Purified anti-human MRGX2
Supplier:  Biolegend 359002
Encompass_Preferred
Purified anti-human MRGX2 [K125H4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-605-355