Learn More
Biolegend Purified anti-human/mouse CD266 (Fn14, TWEAK R)
Catalog No. 50166799
Encompass_Preferred
Purified anti-human/mouse CD266 (Fn14, TWEAK R) [ITEM-4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: FC, ICC, IHC-P, IHC-F, WB; Size: 100 ug
Catalog No. 50-166-799 Supplier Biolegend Supplier No. 314102
Add to Cart
Add to Cart
Purified anti-human/mouse CD266 (Fn14, TWEAK R) [ITEM-4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: FC, ICC, IHC-P, IHC-F, WB; Size: 100 ug