Learn More
Biolegend Purified anti-human/mouse ZAP-70
Supplier:  Biolegend 313402
Encompass_Preferred
Purified anti-human/mouse ZAP-70 [1E7.2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC, WB, IP; Size: 100 ug
Catalog No. 50-166-736