Learn More
Biolegend Purified anti-human NGF
Supplier:  Biolegend 509602
Encompass_Preferred
Purified anti-human NGF [JKhNGF-1]; Isotype: Mouse IgG1; Reactivity: Human, Cross-Reactivity: Cattle (Bovine, Cow), Viper; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-170-065