Learn More
Biolegend Purified anti-human Notch 1
Supplier:  Biolegend 352102
Encompass_Preferred
Purified anti-human Notch 1 [MHN1-519]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, Block; Size: 100 ug
Catalog No. 50-168-693