Learn More
Biolegend Purified anti-human Pro-Major Basic Protein 1 (ProMBP1)
Supplier:  Biolegend 346802
Encompass_Preferred
Purified anti-human Pro-Major Basic Protein 1 (ProMBP1) [J175-7D4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC, IP, IHC-P; Size: 100 ug
Catalog No. 50-168-394