Learn More
Biolegend Purified anti-human TNF-α
Supplier:  Biolegend 502802
Encompass_Preferred
Purified anti-human TNF-α [MAb1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, Neut, WB; Size: 500 ug
Catalog No. 50-169-761