Learn More
Biolegend Purified anti-human TRA-1-60-R
Supplier:  Biolegend 330602
Encompass_Preferred
Purified anti-human TRA-1-60-R [TRA-1-60-R]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC, WB, ICC, IP, RIA; Size: 100 ug
Catalog No. 50-167-582