Learn More
Biolegend Purified anti-human TSLPR (TSLP-R)
Supplier:  Biolegend 322802
Encompass_Preferred
Purified anti-human TSLPR (TSLP-R) [1B4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC, WB; Size: 100 ug
Catalog No. 50-167-139