Learn More
Biolegend Purified anti-human CD198 (CCR8)
Supplier:  Biolegend 360602
Encompass_Preferred
Purified anti-human CD198 (CCR8) [L263G8]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-605-371