Learn More
Biolegend Purified anti-human CD138 (Syndecan-1)
Supplier:  Biolegend 356502
Encompass_Preferred
Purified anti-human CD138 (Syndecan-1) [MI15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC, WB, IHC-P; Size: 100 ug
Catalog No. 50-605-326