Learn More
Biolegend Purified anti-human CD161
Supplier:  Biolegend 307502
Encompass_Preferred
Purified anti-human CD161 [W18070C]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-3336